missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat anti-Mouse Kappa Light Chain Secondary Antibody, PE

Product Code. 10003593
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10003593 1 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10003593 Supplier Invitrogen™ Supplier No. MKAPPA04
  • ONLINE EXCLUSIVE PRICE
    329.00 €
    266.49 € / 1mL
    Save 62.51 €
    19% Off
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Secondary Antibody

Rat anti-Mouse Kappa Light Chain Secondary Antibody for Flow, ELISA

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse Kappa Light Chain
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone 187.1
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with <0.1% sodium azide; pH 7.5
Gene Alias Ig kappa chain, Igk, Ig-kappa, Iglc
Host Species Rat
Immunogen Mouse Kappa light chain.
Purification Method purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.