missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rat Brain Membrane Tissue Lysate (Adult Membrane Normal)

Product Code. 18797053
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18797053 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18797053 Supplier R&D Systems™ Supplier No. NB82060713
  • ONLINE EXCLUSIVE PRICE
    563.00 €
    532.35 € / 0.1mg
    Save 30.65 €
    5% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems.

The Rat Brain Membrane Tissue Lysate (Adult Membrane Normal) is derived from rat. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 0.1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Purity Protein
Test Specificity Membrane Protein - Rat Normal Tissue: Brain
Cellular Fraction Membrane
Tissue Condition Normal
Cross Reactivity Rat
Host Species Rat
Concentration 2 mg/mL
Product Type Rat Brain Membrane Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Product Line Novus
Protein State Native
For Use With (Application) Western Blot
Tissue Nervous
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.