missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat Gamma Globulin
Descrizione
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Storage: After opening, dilute only enough antibody for a single day's use. Country of Origin: USA.
Rat IgG is a type of immunoglobulin G (IgG) antibody that is commonly used in scientific research and diagnostics. It is produced by rats in response to foreign antigens, and is widely used as a tool for detecting and measuring the presence of specific proteins or other molecules in biological samples.
Specifica
Specifica
| Content And Storage | 4°C |
| Form | Liquid |
| Product Type | Rat Gamma Globulin |
| Quantity | 10 mg |
| Concentration | 11.3 mg/mL |
| Formulation | PBS with no preservative; pH 7.6 |
| For Use With (Application) | Control, ChIP Assay, Immunoprecipitation, Flow Cytometry |
| Isotype | Ig |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?