missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG VisUCyte HRP Polymer Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems VC005-025
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Rat samples. It is validated for Immunohistochemistry.
Specifications
| IgG | |
| Polyclonal | |
| Immunohistochemistry | |
| Immunoglobulin G, ImmunoglobulinG | |
| Rat IgG | |
| 25 mL | |
| Secondary | |
| Rat | |
| IgG |
| Immunohistochemistry | |
| HRP | |
| Supplied as a solution in PBS with preservative. | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| 6 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction