Learn More
Rat IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems VC005-050
 View more versions of this product
Product Code. 18797818
  • 522.00 € / 50mL
Description
IgG Polyclonal specifically detects IgG in Rat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG
Polyclonal
Immunohistochemistry
Immunoglobulin G, ImmunoglobulinG
Rat IgG
50 mL (2 x 25 mL)
Secondary
Rat
IgG
Immunohistochemistry
HRP
Supplied as a solution in PBS with preservative.
Goat
Affinity Purified
RUO
3500
6 months from date of receipt, 2 to 8 degreesC as supplied.