missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IL-4 ELISA (For Lysates) Kit

Product Code. 17155043
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 96
This item is not returnable. View return policy

Product Code. 17155043

Brand: Invitrogen™ ERA30RB

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Rat IL-4 quantitates rat IL-4 in cell lysate, tissue homogenate. The assay will exclusively recognize both natural and recombinant rat IL-4.

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number B cell growth factor 1; B
Assay Range 1.5 to 160 pg/mL
Assay Sensitivity 1.5 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Cell Lysates, Tissue Homogenates
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 287287
Gene Symbol Il4
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 50 μL, Tissue Homogenate 50 μL
Storage Requirements 2°C to 8°C
Target IL-4
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.