missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat LXR beta (aa 113-126) Synthetic Peptide

Product Code. 11584890
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 11584890

Brand: Invitrogen™ PEP271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic Peptide

Rat LXR beta (residues 113-126) Synthetic Peptide, PEP-271, from Invitrogen.

PEP-271 is a 14 amino acid synthetic peptide whose sequence corresponds to residues 113-126 of rat LXR beta. The sequence of this peptide is (amino to carboxy terminus): R113-Y-A-C-R-G-S-G-T-C-Q-M-D-A126This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, LXR beta, catalog # PA1-333. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 58851
Species Rat
Purification Method purified
Accession Number Q62755
Common Name Rat LXR beta (aa 113-126)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Nr1h2
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence R113 - Y - A - C - R - G - S - G - T - C - Q - M - D - A126
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.