missing translation for 'onlineSavingsMsg'
Learn More

Rat monoclonal [SB84a] Anti-Mouse IgG2a heavy chain (Alexa Fluor™ 488), Abcam

Product Code. 32116985 Shop All Abcam Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116985 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116985 Supplier Abcam Supplier No. ab172324250ug
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody

Suitable for Flow Cyt, IHC-Fr. Ideal for fluorescent cell and tissue imaging. Cited in 2 publications.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Mouse IgG2a
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone SB84a
Conjugate Alexa Fluor 488
Formulation Preservative: 0.02% Sodium azide, Constituents: PBS, 23% Glycerol (glycerin, glycerine), 1% BSA
Host Species Rat
Immunogen Mouse IgG2a
Purification Method Affinity purification
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -20°C
Product Type Primary Antibody
Form Liquid
Isotype IgG1
Shipping Condition Blue Ice
Target Molecule Mouse IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.