missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ Rat Neonatal Retinal Cells
Click to view available options
Quantity:
≥ 200,000 cells
Unit Size:
Each
Description
- Ready to use.
- High-quality primary cells from Sprague Dawley rats.
- No tissue dissection or animal handling required.
- Increased data reproducibility.
- Characteristic morphology and staining tests confirm tissue origin and cell type.
- Cells are characterized by immunostaining for neuron specific class III β-tubulin (Tuj-1), specific neuronal protein gene product (PGP 9.5), ganglion cell marker, Thy1.1 and GFAP.
- Negative for mycoplasma, bacteria, yeast and fungi.
- Certificate of Analysis (CoA) is available for every batch
- Applications:
- General ophthalmic research
- Posterior segment disease
- Inflammation
- Degeneration
- Neoplasms
- Drug delivery
- Cell therapies
- Toxicology and cytotoxicity
- Gene expression
Specifications
Specifications
| Certifications/Compliance | Certificate of analysis with each lot |
| Content And Storage | Cryopreservation |
| Cell Type | Neonatal Retinal Cells |
| Species | Rat |
| Quantity | ≥ 200,000 cells |
| Source | Sprague Dawley Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction