missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat RAGE (aa 362-380) Synthetic Peptide

Product Code. 11524850
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11524850 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11524850 Supplier Invitrogen™ Supplier No. PEP194
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Synthetic Peptide

RAGE (residues 362-380) Synthetic Peptide, PEP-194, from Invitrogen.

PEP-194 is a 20 amino acid synthetic peptide. The sequence of this peptide is (amino to carboxy terminus): C (362) W R K R Q P R (R/L) E E R K A P E S Q E D (NH2) (380).This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, catalog # PA1-075. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 81722
Species Rat
Purification Method purified
Accession Number Q63495
Common Name Rat RAGE (aa 362-380)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Ager
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO
Sequence C(362) W R K R Q P R (R/L) E E R K A P E S Q E D(NH2)(380)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.