missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Rat ROR1 Protein, His Tag

Product Code. 15827607
Change view
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15827607 1 mg 1mg
15817607 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15827607 Supplier ACROBiosystems Supplier No. RO1R52211MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tyrosine-protein kinase transmembrane receptor ROR1 is also known as Neurotrophic tyrosine kinase, receptor-related 1 (NTRKR1), which belongs to the protein kinase superfamily or tyr protein kinase family or ROR subfamily. ROR1 contains 1 FZ (frizzled) domain, 1 Ig-like C2-type (immunoglobulin-like) domain, 1 kringle domain, 1 protein kinase domain. ROR1 is expressed at high levels during early embryonic development. The expression levels drop strongly around day 16 and there are only very low levels in adult tissues. Isoform Short is strongly expressed in fetal and adult CNS and in a variety of human cancers, including those originating from CNS or PNS neuroectoderm. ROR1 could interact with casein kinase 1 epsilon (CK1ε) to activate phosphoinositide 3-kinase-mediated AKT phosphorylation and cAMP-response-element-binding protein (CREB), which was associated with enhanced tumor-cell growth.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.5 kDa. The protein migrates as 60-67 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number EDL97819
Molecular Weight (g/mol) 43.5 kDa
Quantity 1 mg
Source Rat ROR1, His Tag (RO1-R5221) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Glu 403 (Accession # EDL97819).
Immunogen Gln 30 - Glu 403
Storage Requirements -20°C
Gene Alias ROR1,NTRKR1
Common Name ROR1
Conjugate Unconjugated
Species Rat
Protein Tag His Tag
Buffer PBS, pH7.4
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Bio-Markers and CD Antigens
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.