missing translation for 'onlineSavingsMsg'
Learn More

Megro™ Ratiomed™ Pill Splitter

Product Code. 15972212
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15972212 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15972212 Supplier Megro™ Supplier No. MEG 123060
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Conveniently and easily cut pills into two halves for accurate dosing.

  • Inside blade
  • Simply press down to split pills
  • Can also be used to store pills until needed
  • Convenient size, fits in any pocket

Specifications

For Use With (Application) Pill Splitting
Size 83 x 33 x 25 mm
Color Blue
Quantity 500/Cs.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.