missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals RBC lysis buffer for human

Product Code. 15465269
Change view
Click to view available options
Quantity:
250mL
500mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465269 250mL 250mL
15475269 500mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465269 Supplier Thermo Scientific Chemicals Supplier No. J62990.AK
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Ajouter au panier
Edge
Ajouter au panier
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

For lysis of human red blood cells

For lysis of human red blood cells and can be used to lyse reticulocytes, erythrocytes, and mature RBCs.

RBC Lysis Buffer for human is used in the isolation of deoxyribonucleic acid (DNA) and ribonucleic acid (RNA).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Spécification

Quantity 250mL
Format Liquid
Product Type Cell Lysis Reagent
For Use With (Application) For lysis of human red blood cells.
Content And Storage 2° to 8°C
What are the concentrations of the components of the RBC lysis buffer for human (Cat. No. J62990.xx)?

RBC lysis buffer for human contains 150 mM ammonium chloride, 10 mM potassium bicarbonate, and 0.1 mM EDTA.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.