missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ 10X RBC Lysis Buffer (Multi-species)

Product Code. 15270658
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15270658 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15270658 Supplier Invitrogen™ Supplier No. 00430054
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specially formulated for optimal lysis of erythrocytes in single-cell suspensions of peripheral blood and hematopoietic tissues such as spleen. This buffer can be used for lysis of human, mouse, rat, canine, and non-human primate samples.

Contains ammonium chloride, which lyses red blood cells with a minimal effect on lymphocytes when used as instructed.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
Format Liquid
Product Type Lysis Buffer
Volume (Metric) 50 mL
Product Line eBioscience
compliance-icons
Product Identifier
  • eBioscience RBC Lysis Buffer (Multi-species) 10X
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ammonium chloride

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.