missing translation for 'onlineSavingsMsg'
Learn More

Medline Scientific™ RBS™ Alkaline Detergent

Product Code. 12893512
Change view
Click to view available options
Volume (Metric):
5 L
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
12893512 5 L 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12893512 Supplier Medline Scientific™ Supplier No. RBST105
  • ONLINE EXCLUSIVE PRICE
    119.00 €
    113.80 € / 5 L
    Save 5.20 €
    4% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use this alkaline detergent for manual cleaning, soaking and ultrasonic cleaning of glassware and laboratory equipment.

  • Alkaline Detergent
  • For cleaning lab equipment

Specifications

Product Type Alkaline Detergent
Volume (Metric) 5 L
compliance-icons
Product Identifier
  • RBS T 105
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.