Learn More
Cytiva RC 10 Packing Reservoir
Product Code. 10800891 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
15 mL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
10800891 15 mL Each
1 options
Product Code. 10800891
Brand: Cytiva 19500701
Only null left
Add to basket
Add to basket
Curved walls minimise the separation of matrix beads
RC packing reservoirs, which are available for all three C column types, are quick and easy to assemble. Curved walls minimise the separation of matrix beads by size during packing to produce homogeneous columns. Once the matrix is in the column, the RC reservoir can be connected to a pump or Mariotte flask to give optimal flow rates for column packing. Packing reservoirs for each column diameter optimise packing conditions.
TRUSTED_SUSTAINABILITY
Specifications
Material Acetal Plastic (POM), Polypropylene, Fluroro RubberNitrile Rubber (NBR), Superpolyxymethylene
Type RC 10 Packing Reservoir
Volume (Metric) 15 mL
Quantity 1 Pack