missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Uses solid aluminium synthesis blocks to significantly reduce potential hazards compared to oil or sand baths while simplifying handling and cleaning. Helps reduce costs and make workflows more efficient
- Wireless Temperature and pH Measurement in the Reaction Medium
- Wireless sensor transmits measured values wirelessly to the magnetic stirrer
- An optional pH electrode can be connected via the BNC port, enabling pH values to be displayed directly on the screen without an additional external instrument
- Eliminating sensor cables increases flexibility in setup and reduces risks from damaged or burnt sensor cables
- Increased Safety Through Dual Temperature Monitoring
- Includes the PT 1000.60 wired temperature probe in addition to the wireless sensor, allowing two temperatures to be monitored simultaneously
- The wired probe monitors and limits the temperature of the synthesis block or heat carrier, while the wireless sensor controls the temperature directly in the reaction medium
- Separate temperature measurement significantly increases experiment safety, particularly in the event of glass breakage
- Both temperature values are automatically displayed in parallel on the screen
- Wireless dongle allows connection to a laboratory app for visualising measurement data
- A Well-Designed System Solution for the Synthesis Setup
- Flask holder for 250 mL round-bottom flasks
- Synthesis block made of solid anodised aluminium, temperature-resistant up to 300°C
- Stand rod and support rod for positioning sensors and accessories
- Two cross-sockets
- Universal clamp for secure attachment of the round-bottom flask
- Elliptical magnetic stir bars optimally adapted to the shape of the round-bottom flask
- An additional stand rod can optionally be mounted via the second stand mount for secure attachment of a reflux condenser
Specifications
Specifications
| Top Plate Material | Aluminum Alloy |
| Voltage | 220/230 V |
| Size | 135 mm dia. |
| Temperature (Metric) | 0°C to 310 °C |
| Type | Magnetic Stirrer |
| Plug Type | UK |
| No. of Positions | 1 |
| Max. Container Size | 20 L |
| Stirring Range | 50 to 1500 rpm |
| Warranty | 2 Years |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?