missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Rea-des™ 2000 Cleaning and Disinfecting Agent

Product Code. 10258301
Click to view available options
Container Type:
Bottle
Drum
Unit Size:
1 L
30 L
This item is not returnable. View return policy

Product Code. 10258301

Brand: Hirschmann™ 9730101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For the cleaning of laboratory glass devices and instruments, washable surfaces of all types such as floors, walls, bath tubs, sinks, fittings, toilets, cabinets, doors and more.

  • Cleans and disinfects in a single working operation
  • Bactericide, fungicide, virus inactivating in case of hepatitis B and HIV
  • Non-corrosive in case of common materials
  • Rinses off without leaving residue
  • Economical: 0.8 to 2% working solution
  • For use in all areas with a risk of infection, such as hospitals, doctors’ practices, the pharmaceutical and food processing industry and more
  • Also suitable for senior citizen and nursing homes, schools, kindergartens and large kitchens

Specifications

Concentration 0.8 to 2%
Container Type Bottle
For Use With (Application) Cleaning, disinfecting
For Use With (Equipment) Glass devices, washable surfaces
Volume (Metric) 1 L
compliance-icons compliance-icons
Product Identifier
  • rea-des 2000, liquid cleaning
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P391-Collect spillage.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : trisodium nitrilotriacetate, sodium etasulphate, citric acidmonohydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.