missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Rea-sol™ 30 L Rapid Cleaning Agent

Product Code. 10014061
Click to view available options
Volume (Metric):
30 L
Unit Size:
30 L
This item is not returnable. View return policy

Product Code. 10014061

Brand: Hirschmann™ 9720130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cleans materials such as glass, porcelain, rubber, plastics and metals rapidly, automatically and carefully of oil, grease, wax, resin, silicone, blood, protein and serum residues.

  • With anti-corrosion additives
  • Graduations are not attacked
  • Sensitive tests are not influenced
  • Also suitable for cleaning and decontamination of walls and floors
  • Free of chlorine and caustic alkalis
  • Replacement to a broad extent for chromosulfuric acid
  • Good skin toleration and non-toxic
  • Environmentally friendly as the active ingredients are biologically degradable
  • Extremely economical: 1% working solution, can be used several times
  • Also suitable for use in electronic baths
  • Provided in a 30 L drum

Specifications

Product Type Rapid Cleaning Agent
Concentration 1%
Container Type Drum
For Use With (Application) Cleaning and decontamination
For Use With (Equipment) Glass, porcelain, rubber, plastic, metal instruments and surfaces
Volume (Metric) 30 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.