missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Reaction Vessel, 8 mL, RTF version

Product Code. 15882624 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
8 mL
Unit Size:
8mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15882624 8 mL 8mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.
Product Code. 15882624 missing translation for 'mfr' Applied Biosystems™ missing translation for 'supplierNo' 401571

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
If you would like to order, a signed ATCC agreement is required.

A new style reaction vessel is now available for both the Models 430A and 431A Peptide Synthesizers.

This new vessel uses tabbed filters which are placed on the recessed edge inside the vessel. The caps of the new vessel include an internal O-ring to back up the primary seal formed between the vessel, filter, and the cap. These vessels require changing the side of the vortexer bracket to accommodate the vessel and include a shielded vortexer bearing. These design changes address the known difficulties associated with the assembly of the reaction vessels.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 433 A Peptide Synthesizer
Product Type Reaction Vessel
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Quantity 8 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.