missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ 2.5% Normal Goat Serum (1X)

Product Code. 16607825
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16607825 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16607825 Supplier Invitrogen™ Supplier No. R37624
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF) experiments

  • ReadyProbes 2.5% Normal Goat Serum is a ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF) experiments, usually when a goat secondary antibody is used.
  • Incubating cells/tissue with diluted serum from the same species as the host of the secondary antibody typically reduces non-specific binding.
  • Simply cover the tissue section or cells with a few drops of ReadyProbes 2.5% Normal Goat Serum and incubate for 30–60 minutes at room temperature.
  • Ready Probes 2.5% Normal Goat Serum can also be used to dilute primary and secondary antibodies.
  • Ready Probes 2.5% Normal Goat Serum is provided as a ready-to-use reagent with no further dilution or preparation required.
  • The serum is collected from healthy adult animals and the heat-treated, centrifuged, and filtered. It is then checked for cross reactivity using an appropriate immunoassay.
  • The serum is diluted to 2.5% (v/v) in buffer, with 0.08% sodium azide added as a preservative.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Provided as 50-mL solution at ready-to-use (1X) concentration in a clear plastic dropper bottle.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Species Goat
Form Liquid
Product Type Goat Serum
Quantity 50 mL
Product Line ReadyProbes
Reagent Type Blocking Reagent
Shipping Condition Room Temperature or Wet Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.