missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ Avidin/Biotin Blocking Solution (1X)

Product Code. 17152534
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17152534 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17152534 Supplier Invitrogen™ Supplier No. R37627
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Blocks endogenous biotin, biotin receptors, and avidin binding sites present in many types of tissue, reducing background signal in immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) procedures

  • ReadyProbes Avidin/Biotin Blocking Solution blocks endogenous biotin, biotin receptors, and avidin binding sites present in many types of tissue, reducing background signal in immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) procedures.
  • This two-component solution (an avidin-blocking solution and a biotin-blocking solution) is provided in a ready-to-use format—just add a few drops of the avidin component and incubate for 15 minutes, then wash and repeat with the biotin component.
  • ReadyProbes Avidin/Biotin Blocking Solution is used after the blocking of cells/tissue with standard non-specific blocking solution (e.g., diluted serum), but before the primary antibody step.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Avidin/Biotin Blocking Solution
Quantity 1 Kit
Product Line ReadyProbes
Content And Storage A kit containing 18 mL each ready-to-use (1X) solutions of avidin (component A) and biotin (component B) in two clear LDPE dropper bottles.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
With the ReadyProbes Avidin/Biotin Blocking Solution (1X), why is there a step to add biotin to the sample?

The first step is to bind all endogenous biotinylated entities with an unlabeled avidin. The free D-biotin is then added to bind to all biotin binding sites on this added avidin. The free biotin will be washed away before any other labels are added and all biotin binding sites will be occupied on the unlabeled avidin.

I am labeling cells with a biotinylated primary antibody, followed by an avidin dye conjugate, but I'm getting non-specific mitochondrial labeling, even with an avidin-only control. Why is this?

Mitochondria have endogenous biotin, which is being labeled by the streptavidin. You will need to block the endogenous biotin first, after you fix and permeabilize, but before you block for your primary antibody.

How is ReadyProbes Avidin/Biotin Blocking Solution (1X) different from other endogenous biotin blocking kits?

The only difference between the ReadyProbes Avidin/Biotin Blocking Solution (1X) and other endogenous biotin blocking kits is the ease of use. In the ReadyProbes Avidin/Biotin Blocking Solution (1X), we provide the two components already pre-mixed and ready to use.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.