missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ReadyProbes™ Hydrophobic Barrier Pap Pen

Product Code. 17816781 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 Pens (6 mL Each)
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17816781 2 Pens (6 mL Each) Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17816781 Supplier Thermo Scientific™ Supplier No. R3777
Black Friday Starts Here
Use promo code 27984
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The ReadyProbes Hydrophobic Barrier Pap Pen creates a water-repellent barrier when used on a microscope slide or coverslip.

  • The ReadyProbes Hydrophobic Barrier Pap Pen creates a water-repellent barrier when used on a microscope slide or coverslip
  • By lightly drawing a rectangle or circle around the tissue section or cells on the microscope slide, aqueous incubation reagents can be kept inside the circle or rectangle
  • Save precious reagents by incubating with less volume
  • The pen can also be used to keep reagents separate when more than one tissue section is mounted on a microscope slide
  • The light blue barrier line is heat resistant and compatible with most immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) reagents, including detergent-containing reagents
  • It is insoluble in alcohol and acetone, but can be removed with clearing agent (e.g., xylene)
  • When applied as directed, the nib provided with each pen will create a 3 mm wide line
  • The pen design ensures the nib can move and that the nib is not adhered to the pen itself
  • In some instances, during shipping, the nib may become dislodged and fall out. Simply put the nib back into the neck of the pen and use as per the supplied instructions
  • The pen residue does fluoresce – apply the Hydrophobic Barrier a few mm away from tissue section or cells, so it does not interfere with imaging

How to Use

  • Dab the nib of the pen on a paper towel, to make sure the flow of the hydrophobic fluid is uninterrupted
  • Lightly trace a circle or rectangle around the tissue/cells 2-4 times, with a thick line, while keeping a few millimeters distance from the edge of the tissue/cells
  • Let the line dry for 20-30 minutes at room temperature
  • Repeat the process, if needed
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 Pens (6 mL Each)
For Use With (Application) Immunohistochemistry (IHC), Immunocytochemistry (ICC), Immunofluorescence (IF)
Product Line ReadyProbes
Content And Storage Store at room temperature, away from sunlight, or other heat sources.
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.