missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ReadyProbes™ Mouse-on-Mouse IgG Blocking Solution (30X)

Product Code. 16680534
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16680534 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16680534 Supplier Invitrogen™ Supplier No. R37621
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ReadyProbes Mouse-on-Mouse IgG Blocking Solution blocks endogenous binding when mouse IgG primary antibody is used to label mouse tissue.

ReadyProbes Mouse-on-Mouse IgG Blocking Solution blocks endogenous binding when mouse IgG primary antibody is used to label mouse tissue. Most mouse tissues contain mouse IgG antibodies. When an anti-mouse IgG secondary antibody is used on mouse tissue, the antibody cannot distinguish between the endogenous mouse IgG antibodies and the mouse primary antibody used for target protein detection. ReadyProbes Mouse on Mouse IgG Blocking Solution reduces the binding of endogenous antibodies, resulting in increased specific detection in immunohistochemistry (IHC) and immunofluorescence (IF) applications. This reagent is compatible with both fluorescent and chromogenic IHC procedures.

ReadyProbes Mouse-on-Mouse IgG Blocking Solution is provided at a 30X concentration and is diluted to a 1X working concentration (2 drops in 2.5 mL of PBS or TBS). This solution is not used in place of a non-specific blocking solution (e.g., diluted serum), but is used after the tissue has already been blocked with a non-specific solution and before the primary antibody step. Simply add the 1X solution to the tissue and incubate for 30-60 minutes at room temperature.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Blocking Solution
Quantity 1 mL
Product Line ReadyProbes
Form Liquid
Content And Storage Provided as 1-mL solution at 30 x concentration in a LDPE dropper bottle.

Store (at 2°C to 8°C

When stored as directed, product can be used for 6 months after receipt

.One bottle is sufficient to treat approximately 125-250 tissue sections.

)
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
What is the volume for one drop from the ReadyProbes dropper bottles?

The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.

Can I use ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) on rat tissues?

Due to some cross-reactivity of anti-mouse antibodies to rat antibodies, using ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) on rat tissues may offer some level of blocking, but it may not be sufficient. For rat tissues, ideally you should use a rat-on-rat blocking.

Can I combine the standard blocking step (using serum or BSA) with the ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) to save time?

No. These two blocking steps cannot be combined. Doing so would result in inadequate blocking for both non-specific entities and mouse antibodies.

Do I need to use ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) on cultured mouse cells?

Assuming that the cultured mouse cells are not expressing mouse antibodies, ReadyProbes Mouse-on-Mouse IgG Blocking Solution (30X) is not needed for cultured mouse cells.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.