missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Reagent Reservoir Chiller

Product Code. 16340412 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Volume (Metric):
100 mL
50 and 55 mL
Color:
Blue
Purple
Purple & Blue
Unit Size:
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Color Unit Size
16340412 50 and 55 mL Blue Pack of 2
16330412 50 and 55 mL Purple Pack of 2
16350412 50 and 55 mL Purple & Blue Pack of 2
16360412 100 mL Purple Pack of 2
16370412 100 mL Blue Pack of 2
16380412 100 mL Purple & Blue Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16340412
  • ONLINE EXCLUSIVE PRICE
    111.00 €
    104.05 € / Pack of 2
    Save 6.95 €
    6% Off
View all promotions
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

The Reservoir Chiller is an ice-free, non-toxic way to extend working period and preserve integrity for temperature sensitive reagents.

  • Reagents are kept at less than 5°C (41°F) for approximately 3 hours
  • Ice-free solution eliminates the risk of contamination from messy ice baths
  • Stackable, save space when storing in the freezer or on the bench top
  • Symmetrical cooler design allows reagent reservoirs to be mounted in either direction
  • Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
  • Made from sonic welded, engineered polypropylene that ensures a leak free design
  • Not intended for autoclave, do not use with liquid nitrogen
  • Reusable
  • Two-year warranty

Specifications

Product Type Reservoir Chiller
Autoclavable Autoclavable
Color Blue
Description Reagent Reservoir Chiller
Device Holds Reagent Reservoirs
Material Polypropylene
Volume (Metric) 50 and 55 mL
For Use With (Application) Reagent
For Use With (Equipment) Reagent Reservoirs
Includes Chiller
Model 120731F
Quantity 1
Shape Rectangular
Warranty 2 Years
Dimensions (L x W) 5.6 x 2.9 in.
Dimensions (L x W x H) 5.6 x 2.9 x 1.75 in.
Length (English) 5.6 in.
Length (Metric) 14.4 cm
Width (English) 5.70 in.
Width (Metric) 14.30 cm
Height (English) 1.75 in.
Height (Metric) 4.5 cm
Weight (English) 0.31 lb.
Weight (Metric) 0.14 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.