missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Reagent Vessel, 60 mL

Product Code. 12460030 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Konelab Clinical Chemistry Analyzers
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12460030 Konelab Clinical Chemistry Analyzers Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12460030 Supplier Thermo Scientific™ Supplier No. 981455
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Plastic reagent vessels for use with biochemical analyzers.

Plastic sample containers with a volume of 60 ml, suitable for biochemical analyzers of the Konelab series.

  • Store at room temperature
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Photometric Analysis
For Use With (Equipment) Konelab Clinical Chemistry Analyzers
Capacity (Metric) 60 mL
Quantity 12 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.