missing translation for 'onlineSavingsMsg'
Learn More

LAL Reagent Water, Lonza™

Product Code. 11620451
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11620451 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11620451 Supplier Lonza Supplier No. W50100
  • ONLINE EXCLUSIVE PRICE
    38.99 €
    30.51 € / 100mL
    Save 8.48 €
    21% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Routinely monitored and tested to meet all USP requirements for WFI. Lonza™ LAL Reagent Water is equivalent to Water for Bacterial Endotoxins Test (BET).

Tested to ensure:
  • Endotoxin levels are < 0.005 EU/mL
  • Positive Product Control (PPC) requirement for the LAL Reagent Water of 75-150%
  • Endotoxin free
  • Equivalent to Water for Bacterial Endotoxins Test (BET).
TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5
Color Clear, Colorless
Melting Point 0.1°C
Boiling Point 100°C
Molecular Formula H2O
Endotoxin <0.005 EU/mL
Solubility Information Soluble
Molecular Weight (g/mol) 18.01
Grade BET, WFI
Odor Odorless
Packaging Plastic (PETG) bottle with HDPE screw cap
Vapor Pressure 23 mbar / 17.5 hPa at 20°C
Density 1g/cm3
Quantity 100 mL
Physical Form Liquid
Chemical Name or Material Water
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.