missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Reboxetine mesylate

Product Code. 30001365
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001365 10 mg 10mg
18035482 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001365 Supplier R&D Systems™ Supplier No. 1982/10
  • ONLINE EXCLUSIVE PRICE
    316.14 €
    283.50 € / 10mg
    Save 32.64 €
    10% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective noradrenalin uptake inhibitor; orally active

Potent and selective inhibitor of noradrenalin uptake (Ki values are 1.1, 129 and > 10000 nM for rat NET, SERT and DAT respectively). Displays > 1000-fold selectivity over α-adrenoceptors, 5-HT, dopamine and muscarinic ACh receptors. Orally active antidepressant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98769-84-7
Molecular Formula C20H27NO6S

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 409.5
Purity >99%
Molecular Weight (g/mol) 409.497
Chemical Name or Material Reboxetine mesylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.