missing translation for 'onlineSavingsMsg'
Learn More

Reboxetine Mesylate, TRC

Product Code. 30137619
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137619 25 mg 25mg
18022020 5 mg 5mg
18072410 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137619 Supplier Toronto Research Chemicals Supplier No. TRCR14200025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98769-84-7
Formula Weight 409.1559
InChI Formula InChI=1S/C19H23NO3.CH4O3S/c1-2-21-16-10-6-7-11-17(16)23-19(15-8-4-3-5-9-15)18-14-20-12-13-22-18;1-5(2,3)4/h3-11,18-20H,2,12-14H2,1H3;1H3,(H,2,3,4)/t18-,19-;/m1./s1
IUPAC Name (2R)-2-[(R)-(2-ethoxyphenoxy)-phenylmethyl]morpholine;methanesulfonic acid
Molecular Formula C19 H23 N O3 . C H4 O3 S
Molecular Weight (g/mol) 409.5
SMILES CCOc1ccccc1O[C@@H]([C@H]2CNCCO2)c3ccccc3.CS(=O)(=O)O
Synonym Reboxetine Mesilate, Morpholine, 2-[(2-ethoxyphenoxy)phenylmethyl]-, (R*,R*)-(±)-, methanesulfonate, Morpholine, 2-[(R)-(2-ethoxyphenoxy)phenylmethyl]-, (2R)-rel-, methanesulfonate (9CI), (±) Reboxetine mesylate, Davedax, Edronax, FCE 20124, PNU 155905E, PNU 155950E, Reboxetine mesilate, Reboxetine mesylate, Morpholine, 2-[(R)-(2-ethoxyphenoxy)phenylmethyl]-, (2R)-rel-, methanesulfonate (1:1)
Chemical Name or Material Reboxetine Mesilate

Specifications

Quantity 25 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.