missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rec Protein A-HRP

Product Code. 10581665
Change view
Click to view available options
Quantity:
3 mg
Unit Size:
3mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10581665 3 mg 3mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10581665 Supplier Invitrogen™ Supplier No. 101123
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rec Protein A-HRP

This recombinant protein A is purified from cytosol of an E. coli strain expressing the genes coding for Staphylococcus aureus protein A. This version is conjugated with highly purified horseradish peroxidase.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate HRP
Quantity 3 mg
Form Solution
Protein Subtype Protein A
Content And Storage 2-8°C. Do Not Freeze.
Shipping Condition Wet Ice
compliance-icons
Product Identifier
  • Rec Protein A-HRP
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: 5-chloro-2-methylisothiazol -3(2H)-one,2-methylisothiazol-3(2H)-o ne

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.