missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Rechargeable Lithium-ion Battery for M510/M610/M710 Printers

Product Code. 18724614 Shop All Brady Products
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18724614 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18724614 Supplier Brady™ Supplier No. 175673
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Work longer with this Li-ION M510, M610 and M710 battery pack. It enables cordless printer operation so you can print labels where and when you need them.

  • Removable, rechargeable Li-ION battery pack
  • Battery can be charged either while installed in an M610 or M710 printer, or while removed
  • Prints up to 4,500 labels per charge

Specifications

Product Type Battery
For Use With (Equipment) BradyPrinter M610, Brady M710 Label Printer
Width (Metric) 59.94 mm
Height (English) 3.72 in.
Height (Metric) 94.50 mm
Description Rechargeable Lithium-ion Battery
Includes Li-Ion Battery Pack
For Use With (Application) Asset Tracking, Circuit Board Labelling, Data and Telecommunications Labelling, Electrical Labelling, General Identification, Wire and Cable Labelling, Facility Identification, Safety Identification
Quantity 1 Ea.
Alternate Part Number M-LION-BATTERY
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.