missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bacteriophage Shiga-like toxin 1 subunit B Protein

A DNA sequence encoding the Enterobacteria phage H19B (Bacteriophage H19B) Shiga-like toxin 1 subunit B, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6941-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16016790

  • 940.00 € / 100µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Shiga-like toxin 1 subunit B Protein
Research Use Only
stxB
Recombinant Protein
E. coli
TPDCVTGKVE YTKYNDDDTF TVKVGDKELF TNRWNLQSLL LSAQITGMTV TIKTNACHNG GGFSEVIFR
Greater than 90% as determined by SDS-PAGE.
100 μg
Not determined.
Full Length
Bacteriophage
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions