missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine aFGF Protein

Product Code. 15934088
Click to view available options
Quantity:
1 μg
100 μg
5 μg
Unit Size:
100µg
1µg
5µg
This item is not returnable. View return policy

Product Code. 15934088

Brand: enQuireBio™ QP105081ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the aFGF was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 281160
Name aFGF Protein
Quantity 1 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol FGF1
Biological Activity Stimulates growth of bovine capillary endothelial cells by 3-5 fold over 5% calf serum at 10-25ng/ml FGF and 0ug/ml heparin.
Product Type Recombinant Protein
Cross Reactivity Bovine
Species Bovine
Protein Tag Untagged
Sequence MAEGETTTFT ALTEKFNLPL GNYKKPKLLY CSNGGYFLRI LPDGTVDGTK DRSDQHIQLQ LCAESIGEVY IKSTETGQFL AMDTDGLLYG SQTPNEECLF LERLEENHYN TYISKKHAEK HWFVGLKKNG RSKLGPRTHF GQKAILFLPL PVSSD
Buffer Each 5microg aFGF were lyophilized from 0.5ml solution containing 1mM sodium phosphate, pH 7 after filtration over a low binding membrane.
Purity or Quality Grade Greater than 90%.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.