missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Agouti-signaling Protein
A DNA sequence encoding the Bos taurus (Bovine) Agouti-signaling protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7635-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_996674.1 | |
Agouti-signaling Protein | |
1 mg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
404192 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ASIP | |
Recombinant Protein | |
E. coli | |
HLAPEEKPRD ERNLKNNSSM NLLDFPSVSI VALNKKSKKI SRNEAEKKKR PSKRKAPMKN VARTRPPPPT PCVATRDSCK PPAPACCDPC AFCQCRFFRS ACSCRVLNPT C |