missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Alkaline Phosphatase Protein

Product Code. p-7148319
Click to view available options
Quantity:
1 mg
5 mg
30 mg
Unit Size:
1mg
30mg
5mg
This item is not returnable. View return policy

Product Code. 15924118

Brand: enQuireBio™ QP105181mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Alkaline Phosphatase was constructed and used to recombinantly synthesize the protein.

Specifications

Name Alkaline Phosphatase Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity >1000 U/mg (pH 9.6, 25C and 0.025M glycin, 10% glycerol as buffer.
Product Type Recombinant Protein
Cross Reactivity Bovine
Species Bovine
Protein Tag Untagged
Sequence MQGACVLLLL GLHLQLSLGL VPVEEEDPAF WNRQAAQALD VAKKLQPIQT AAKNVILFLG DGMGVPTVTA TRILKGQMNG KLGPETPLAM DQFPYVALSK TYNVDRQVPD SAGTATAYLC GVKGNYRTIG VSAAARYNQC KTTRGNEVTS VMNRAKKAGK SVGVVTTTRV QHASPAGAYA HTVNRNWYSD ADLPADAQMN GCQDIAAQLV NNMDIDVILG GGRKYMFPVG TPDPEYPDDA SVNGVRKRKQ NLVQAWQAKH QGAQYVWNRT ALLQAADDSS VTHLMGLFEP ADMKYNVQQD HTKDPTLQEM TEVALRVVSR NPRGFYLFVE GGRIDHGHHD DKAYMALTEA GMFDNAIAKA NELTSELDTL ILVTADHSHV FSFGGYTLRG TSIFGLAPSK ALDSKSYTSI LYGNGPGYAL GGGSRPDVND STSEDPSYQQ QAAVPQASET HGGEDVAVFA RGPQAHLVHG VEEETFVAHI MAFAGCVEPY TDCNLPAPTT ATSIPDAAHL AASPPPLALL AGAMLLLLAP TLY
Buffer 50% glycerol, 5mM MgCl2, 0.1mM ZnCl2 and 5mM TRIS, pH 7.0.
Purity or Quality Grade 95% pure by Gel Filtration.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.