missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Alkaline Phosphatase Protein
Click to view available options
Quantity:
1 mg
5 mg
30 mg
Unit Size:
1mg
30mg
5mg
Specifications
Specifications
| Name | Alkaline Phosphatase Protein |
| Quantity | 30 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Biological Activity | >1000 U/mg (pH 9.6, 25C and 0.025M glycin, 10% glycerol as buffer. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Bovine |
| Species | Bovine |
| Protein Tag | Untagged |
| Sequence | MQGACVLLLL GLHLQLSLGL VPVEEEDPAF WNRQAAQALD VAKKLQPIQT AAKNVILFLG DGMGVPTVTA TRILKGQMNG KLGPETPLAM DQFPYVALSK TYNVDRQVPD SAGTATAYLC GVKGNYRTIG VSAAARYNQC KTTRGNEVTS VMNRAKKAGK SVGVVTTTRV QHASPAGAYA HTVNRNWYSD ADLPADAQMN GCQDIAAQLV NNMDIDVILG GGRKYMFPVG TPDPEYPDDA SVNGVRKRKQ NLVQAWQAKH QGAQYVWNRT ALLQAADDSS VTHLMGLFEP ADMKYNVQQD HTKDPTLQEM TEVALRVVSR NPRGFYLFVE GGRIDHGHHD DKAYMALTEA GMFDNAIAKA NELTSELDTL ILVTADHSHV FSFGGYTLRG TSIFGLAPSK ALDSKSYTSI LYGNGPGYAL GGGSRPDVND STSEDPSYQQ QAAVPQASET HGGEDVAVFA RGPQAHLVHG VEEETFVAHI MAFAGCVEPY TDCNLPAPTT ATSIPDAAHL AASPPPLALL AGAMLLLLAP TLY |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction