missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine DNase Protein
A cDNA sequence encoding the DNase was constructed and used to recombinantly synthesize the protein.
136.00 € - 1015.00 €
Specifications
Gene ID (Entrez) | 282217 |
---|---|
Name | DNase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | DNASE1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15998058
|
enQuireBio™
QP11693-20MG |
20 mg |
136.00 €
20mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978058
|
enQuireBio™
QP11693-100MG |
100 mg |
203.00 €
100mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15988058
|
enQuireBio™
QP11693-1GRAM |
1 g |
1015.00 €
1g |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
282217 | |
Research Use Only | |
DNASE1 | |
Bovine | |
Untagged |
DNase Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Bovine | |
MRGTRLMGLL LALAGLLQLG LSLKIAAFNI RTFGETKMSN ATLASYIVRI VRRYDIVLIQ EVRDSHLVAV GKLLDYLNQD DPNTYHYVVS EPLGRNSYKE RYLFLFRPNK VSVLDTYQYD DGCESCGNDS FSREPAVVKF SSHSTKVKEF AIVALHSAPS DAVAEINSLY DVYLDVQQKW HLNDVMLMGD FNADCSYVTS SQWSSIRLRT SSTFQWLIPD SADTTATSTN CAYDRIVVAG SLLQSSVVPG SAAPFDFQAA YGLSNEMALA ISDHYPVEVT LT |