missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine ECGS Protein
A cDNA sequence encoding the ECGS was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11736-100mg
Additional Details : Weight : 0.01000kg
Specifications
ECGS Protein | |
Research Use Only | |
Recombinant Protein | |
Bovine | |
The Endothelial-Mitogen was lyophilized from a concentrated solution containing 0.1M NaCl and 5mM NaH2PO4, pH 7.4. |
100 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Bovine | |
Untagged |