missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine ECGS Protein

A cDNA sequence encoding the ECGS was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11736-5mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15988188

  • 131.00 € / 5mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

ECGS Protein
Research Use Only
Recombinant Protein
Bovine
The Endothelial-Mitogen was lyophilized from a concentrated solution containing 0.1M NaCl and 5mM NaH2PO4, pH 7.4.
5 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Bovine
Untagged
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers