missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Enterokinase Protein

A cDNA sequence encoding the Enterokinase was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11793-EC-100IU

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15958388

  • 520.00 € / 100 units
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Enterokinase Protein
Research Use Only
Recombinant Protein
E. coli
MGSKRSVPSR HRSLTTYEVM FAVLFVILVA LCAGLIAVSW LSIQGSVKDA AFGKSHEARG TLKIISGATY NPHLQDKLSV DFKVLAFDIQ QMIDDIFQSS NLKNEYKNSR VLQFENGSII VIFDLLFDQW VSDKNVKEEL IQGIEANKSS QLVTFHIDLN SIDITASLEN FSTISPATTS EKLTTSIPLA TPGNVSIECP PDSRLCADAL KCIAIDLFCD GELNCPDGSD EDNKTCATAC DGRFLLTGSS GSFEALHYPK PSNNTSAVCR WIIRVNQGLS IQLNFDYFNT YYADVLNIYE GMGSSKILRA SLWSNNPGII RIFSNQVTAT FLIQSDESDY IGFKVTYTAF NSKELNNYEK INCNFEDGFC FWIQDLNDDN EWERTQGSTF PPSTGPTFDH TFGNESGFYI STPTGPGGRR ERVGLLTLPL DPTPEQACLS FWYYMYGENV YKLSINISSD QNMEKTIFQK EGNYGQNWNY GQVTLNETVE FKVSFYGFKN QILSDIALDD ISLTYGICNV SVYPEPTLVP TPPPELPTDC GGPHDLWEPN TTFTSINFPN SYPNQAFCIW NLNAQKGKNI QLHFQEFDLE NIADVVEIRD GEGDDSLFLA VYTGPGPVND VFSTTNRMTV LFITDNMLAK QGFKANFTTG YGLGIPEPCK EDNFQCKDGE CIPLVNLCDG FPHCKDGSDE AHCVRLFNGT TDSSGLVQFR IQSIWHVACA ENWTTQISDD VCQLLGLGTG NSSVPTFSTG GGPYVNLNTA PNGSLILTPS QQCLEDSLIL LQCNYKSCGK KLVTQEVSPK IVGGSDSREG AWPWVVALYF DDQQVCGASL VSRDWLVSAA HCVYGRNMEP SKWKAVLGLH MASNLTSPQI ETRLIDQIVI NPHYNKRRKN NDIAMMHLEM KVNYTDYIQP
Greater than 95.0% as determined by SDS-PAGE.
100 IU
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Bovine
Untagged
Bovine EK in 50mM Tris-HCl, pH 8.0, 0.5M NaCl and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers