missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine FGF 2 Protein

Product Code. 15974368
Click to view available options
Quantity:
10 μg
2 μg
Unit Size:
10µg
2µg
This item is not returnable. View return policy

Product Code. 15974368

Brand: enQuireBio™ QP105992ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the FGF 2 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 281161
Name FGF 2 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol FGF2
Biological Activity The recommended concentration in responsive cells is 0.1 to 2ng/ml.
Product Type Recombinant Protein
Cross Reactivity Bovine
Species Bovine
Protein Tag Untagged
Sequence MAAGSITTLP ALPEDGGSGA FPPGHFKDPK RLYCKNGGFF LRIHPDGRVD GVREKSDPHI KLQLQAEERG VVSIKGVCAN RYLAMKEDGR LLASKCVTDE CFFFERLESN NYNTYRSRKY SSWYVALKRT GQYKLGPKTG PGQKAILFLP MSAKS
Buffer The FGF-basic Bovine was lyophilized from a concentrated sterile solution containing 50mM Na2HPO4, pH 7.5 and 0.5% HSA.
Purity or Quality Grade Greater than 95.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.