missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine FGF 21 Protein

Product Code. 15928648
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
This item is not returnable. View return policy

Product Code. 15928648

Brand: enQuireBio™ QP118702ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the FGF 21 was constructed and used to recombinantly synthesize the protein.

Specifications

Name FGF 21 Protein
Quantity 2 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Bovine
Species E. coli
Protein Tag Untagged
Buffer The protein was lyophilized from a concentrated (0.8 mg/ml) solution with 0.4 mg/ml of NaHCO3, pH 8.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by Gel Filtration. (b) Analysis by SDS-PAGE.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.