missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Galectin-1 / LGALS1 Protein
A DNA sequence encoding the Bos taurus (Bovine) Galectin-1 / LGALS1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6293-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_786976.1 | |
Galectin-1 / LGALS1 Protein | |
100 μg | |
Not determined. | |
Full Length | |
Bovine | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
326598 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LGALS1 | |
Recombinant Protein | |
E. coli | |
ACGLVASNLN LKPGECLRVR GEVAADAKSF LLNLGKDDNN LCLHFNPRFN AHGDVNTIVC NSKDAGAWGA EQRESAFPFQ PGSVVEVCIS FNQTDLTIKL PDGYEFKFPN RLNLEAINYL SAGGDFKIKC VAFE |