missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine LH-beta Protein
A DNA sequence encoding the Bos taurus (Bovine) LH-beta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6303-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_776355.1 | |
LH-beta Protein | |
1 mg | |
Not determined. | |
Full Length | |
Bovine | |
His | |
Tris-based buffer, 50% glycerol |
280839 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
LHB | |
Recombinant Protein | |
E. coli | |
SRGPLRPLCQ PINATLAAEK EACPVCITFT TSICAGYCPS MKRVLPVILP PMPQRVCTYH ELRFASVRLP GCPPGVDPMV SFPVALSCHC GPCRLSSTDC GGPRTQPLAC DHPPLPDILF L |