missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine PI3Ka Protein

Product Code. 15952349
Click to view available options
Quantity:
1 μg
2.5 μg
5 μg
Unit Size:
1µg
2.5µg
5µg
This item is not returnable. View return policy

Product Code. 15952349

Brand: enQuireBio™ QP130491ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PI3Ka was constructed and used to recombinantly synthesize the protein.

Specifications

Name PI3Ka Protein
Quantity 1 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Bovine
Species Baculovirus
Protein Tag Untagged
Buffer PI3Ka solution in 25mM HEPES, pH 8.0, 25mM NaCl, 2.5mM MgCl2 and 50% glycerol.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.