Learn More
Invitrogen™ Recombinant Bovine Serum Albumin (rBSA), AOF
Description
Invitrogen Recombinant Bovine Serum Albumin (rBSA), AOF, is designed to deliver exceptional quality and consistent performance for a variety of molecular biology applications that require animal origin-free formulations. Recombinant BSA manufacturing technology helps ensure simplified logistics, regulatory compliance, and protection from animal related illnesses.
Recombinant BSA is a protein used to stabilize enzymes during long-term storage and prolonged enzymatic reactions. The presence of rBSA can enhance enzymatic efficiency in the presence of inhibitors (such as during PCR) and prevent the adhesion of enzymes to reaction tubes and tip surfaces.
Features of Invitrogen rBSA, AOF, include:
- Expressed in yeast—helps ensure correct protein folding and stability
- Animal origin-free (AOF)—suitable for applications requiring non-animal derived components
- RNase, DNase, and protease-free—helps prevent degradation of nucleic acids and proteins
Applications of Invitrogen rBSA, AOF, include:
- Enzyme stabilization—helps maintain enzyme activity during storage and prolonged enzymatic reactions
- PCR and RT-qPCR—helps protects DNA polymerases and reverse transcriptases from sample-based inhibition, resulting in enhanced performance
- DNA digestion with restriction endonucleases—helps ensure enzyme stability during restriction digestion reaction
- ELISA—helps prevent non-specific binding and improve assay specificity
Specifications
Specifications
| Concentration | 20 mg/mL |
| Purity | ≥95% |
| Protein | Bovine Serum Albumin |
| Sensitive to Heat Inactivation | Yes |
| Source | Pichia pastoris |
| Quantity | 5 mg |
| Product Type | Recombinant BSA |
| Research Category | Traditional Cloning, Scarless Cloning, PCR |
| Form | Solution |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.