missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Bovine Thrombin Protein
A cDNA sequence encoding the Thrombin was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10879-250U
Additional Details : Weight : 0.01000kg
Specifications
Thrombin Protein | |
Research Use Only | |
2000 NIH U/mg. | |
Bovine | |
Untagged |
250 U | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
Bovine | |
Lyophilized freeze dry powder formulated with Tris buffer and sodium chloride. |