missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine TROVE2 Protein

A cDNA sequence encoding the TROVE2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13820-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934809

  • 203.00 € / 10µg
Estimated Shipment: 22-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TROVE2 Protein
Research Use Only
Recombinant Protein
Bovine
TROVE2 is supplied in 16mM HEPES buffer pH 8.0, 400mM NaCl, and 20% glycerol.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Bovine
Untagged
Greater than 60% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers