missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis omcA Protein
A DNA sequence encoding the Chlamydia trachomatis omcA, was expressed in the hosts and tags indicated.
421.00 € - 3590.00 €
Specifications
Accession Number | NP_219956.1 |
---|---|
Gene ID (Entrez) | 884216 |
Name | omcA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16037060
|
enQuireBio™
QP7018-10UG |
10 μg |
421.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16077060
|
enQuireBio™
QP7018-50UG |
50 μg |
605.00 €
50µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027060
|
enQuireBio™
QP7018-100UG |
100 μg |
1025.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16057060
|
enQuireBio™
QP7018-200UG |
200 μg |
1690.00 €
200µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16067060
|
enQuireBio™
QP7018-500UG |
500 μg |
2260.00 €
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16047060
|
enQuireBio™
QP7018-1MG |
1 mg |
3590.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_219956.1 | |
omcA Protein | |
Not determined. | |
Full Length | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
884216 | |
Research Use Only | |
omcA | |
Recombinant Protein | |
E. coli | |
CCRIVDCCFE DPCAPIQCSP CESKKKDVDG GCNSCNGYVP ACKPCGGDTH QDAKHGPQAR GIPVDGKCRQ | |
Greater than 90% as determined by SDS-PAGE. |