missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis Probable outer membrane protein PmpD Protein
A DNA sequence encoding the Chlamydia trachomatis Probable outer membrane protein PmpD, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7502-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_220332.1 | |
Probable outer membrane protein PmpD Protein | |
100 μg | |
Not determined. | |
Full Length | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
884611 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
pmpD | |
Recombinant Protein | |
E. coli | |
EFDYSTNVWG FAFGGFRTLS AENLVAIDGY KGAYGGASAG VDIQLMEDFV LGVSGAAFLG KMDSQKFDAE VSRKGVVGSV YTGFLAGSWF FKGQYSLGET QNDMKTRYGV LGESSASWTS RGVLADALVE YRSLVGPVRP TFYALHFNPY VEVSYASMKF PGFTEQGREA RSFEDASLTN ITIPLGMKFE LAFIKGQFSE VNSLGISYAW EAYRKVEGGA VQLLEAGFDW EGAPMDLPRQ ELRVALENNT EWSSYFSTVL GLTAFCGGFT STDSKLGYEA NTGLRLIF |